Anti-Aldolase Rabbit Monoclonal Antibody

Lieferant: Abcam
AB150396-100
ABCAAB150396-100EA 875 EUR
ABCAAB150396-100
Anti-Aldolase Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Aldolase
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR