Anti-EIF5A2 Rabbit Monoclonal Antibody

Lieferant: Abcam
AB150403-100
ABCAAB150403-100EA 875 EUR
ABCAAB150403-100
Anti-EIF5A2 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: EIF5A2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR