Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Lieferant: Biotium
20030 20030-1
20030.EA 289 EUR
20030. 20030-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse