Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Lieferant: Biotium
20034 20034-1
20034.EA 347 EUR
20034. 20034-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken