Anti-IgG Goat Polyclonal Antibody (CF™ 405S)

Lieferant: Biotium

20080 20080-1
20080.EA 289 EUR
20080. 20080-1
Anti-IgG Goat Polyclonal Antibody (CF™ 405S)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® CF405S Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR