Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Lieferant: Biotium

20127 20127-1
20127.EA 347 EUR
20127. 20127-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR