Anti-IgG Donkey Polyclonal Antibody (CF™ 594)

Lieferant: Biotium

20152 20152-1
20152.EA 347 EUR
20152. 20152-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 594)
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR