Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Lieferant: Biotium

20155 20155-1
20155.EA 347 EUR
20155. 20155-1
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR