Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)

Lieferant: Biotium

20158 20158-1
20158.EA 347 EUR
20158. 20158-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR