Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)

Lieferant: Biotium

20174 20174-1
20174.EA 347 EUR
20174. 20174-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Antikörper
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR