Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)

Lieferant: Biotium

20179 20179-1
20179.EA 347 EUR
20179. 20179-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR