Anti-GFP Mouse Monoclonal Antibody (CF®770)

Lieferant: Biotium

20220
20220.EA 462 EUR
20220.
Anti-GFP Mouse Monoclonal Antibody (CF®770)
Antikörper
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: monoclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR