Anti-IgG1 Goat Polyclonal Antibody (CF™ 680)

Lieferant: Biotium

20253 20253-1
20253.EA 404 EUR
20253. 20253-1
Anti-IgG1 Goat Polyclonal Antibody (CF™ 680)
Antikörper
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR