Anti-CD3 Mouse Monoclonal Antibody [clone: CA-3]

Lieferant: BosterBio
MA1015
BSBTMA1015EA 550 EUR
BSBTMA1015
Anti-CD3 Mouse Monoclonal Antibody [clone: CA-3]
Antikörper
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR