Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]

Lieferant: BosterBio
MA1019
BSBTMA1019EA 550 EUR
BSBTMA1019
Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]
Antikörper
Type: Primary
Antigen: CDC6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR