Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]

Lieferant: Boster
BSBTMA1057EA 550 EUR
BSBTMA1057
Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]
Antikörper
Mouse IgG monoclonal antibody for MAP2, microtubule-associated protein 2 (MAP2) detection. Tested with WB, IHC-P in Human;mouse;rat. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR