Anti-CD62L Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1359
BSBTPA1359EA 550 EUR
BSBTPA1359
Anti-CD62L Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for L-selectin(SELL) detection. Tested with WB in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD62L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR