Anti-Lipoamide Dehydrogenase Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1463
BSBTPA1463EA 550 EUR
BSBTPA1463
Anti-Lipoamide Dehydrogenase Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Dihydrolipoyl dehydrogenase, mitochondrial(DLD) detection. Tested with WB, IHC-P; IHC-F; ICC in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Lipoamide Dehydrogenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR