Anti-OVA Mouse Monoclonal Antibody [clone: 3G2E1D9,3D5F4A8,3G1E1D9]

Lieferant: Thermo Fisher Scientific
PIEAMA515307EA 0 EUR
PIEAMA515307
Anti-OVA Mouse Monoclonal Antibody [clone: 3G2E1D9,3D5F4A8,3G1E1D9]
Antikörper
MA5-15307 targets OVA in WB applications and shows reactivity with Tag samples. The MA5-15307 immunogen is ovalbumin. Ovalbumin (OVA) is used in variety of laboratories products which contain OVA as a carrier protein. To raise antibodies against small non-immunogenic molecules,called haptens,it is often necessary to couple them to carrier protein,such as OVA.This antibody is a good reagent that may be used for the development of detection assay in ELISA or western blot to test the remaning OVA for clinical products in Vivo or research products In Vitro.

Type: Primary
Antigen: OVA
Clonality: Monoclonal
Clone: 3G2E1D9,3D5F4A8,3G1E1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR