Anti-APOM Mouse Monoclonal Antibody [clone: 10C3G5]

Lieferant: Thermo Fisher Scientific
PIEAMA515323EA 0 EUR
PIEAMA515323
Anti-APOM Mouse Monoclonal Antibody [clone: 10C3G5]
Antikörper
MA5-15323 targets ApoM in ICC and WB applications and shows reactivity with Human samples. The MA5-15323 immunogen is purified recombinant fragment of ApoM expressed in E. Coli. MA5-15323 detects ApoM which has a predicted molecular weight of approximately 21kDa. The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.

Type: Primary
Antigen: APOM
Clonality: Monoclonal
Clone: 10C3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Primates, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR