MA5-15326 targets S100A10/P11 in IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15326 immunogen is purified recombinant fragment of human P11 expressed in E. Coli. MA5-15326 detects S100A10/P11 which has a predicted molecular weight of approximately 11kDa. The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in exocytosis and endocytosis.
Type: Primary
Antigen: S100A10
Clonality: Monoclonal
Clone: 4E7E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human