Anti-CD34 Mouse Monoclonal Antibody [clone: 4H5E7, 9B10D4]

Lieferant: Thermo Fisher Scientific
PIEAMA515331EA 0 EUR
PIEAMA515331
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H5E7, 9B10D4]
Antikörper
MA5-15331 targets CD34 in IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15331 immunogen is purified recombinant fragment of CD34 expressed in E. Coli. CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110 kD that is selectively expressed on human hematopoietic progenitor cells.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H5E7, 9B10D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR