Anti-CD31 Mouse Monoclonal Antibody [clone: 2F7B2]

Lieferant: Thermo Fisher Scientific
PIEAMA515336EA 0 EUR
PIEAMA515336
Anti-CD31 Mouse Monoclonal Antibody [clone: 2F7B2]
Antikörper
MA5-15336 targets CD31 in IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15336 immunogen is purified recombinant fragment of human CD31 expressed in E. Coli. The protein encoded by this gene is found on the surface of platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions. The encoded protein is a member of the immunoglobulin superfamily and is likely involved in leukocyte migration, angiogenesis, and integrin activation.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 2F7B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR