Anti-BTK Mouse Monoclonal Antibody [clone: 7F12H4]

Lieferant: Thermo Fisher Scientific
PIEAMA515337EA 0 EUR
PIEAMA515337
Anti-BTK Mouse Monoclonal Antibody [clone: 7F12H4]
Antikörper
MA5-15337 targets BTK in IF, IHC, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15337 immunogen is purified recombinant fragment of BTK expressed in E. Coli. MA5-15337 detects BTK which has a predicted molecular weight of approximately 77kDa. The protein encoded by this gene plays a crucial role in B-cell development. Mutations in this gene cause X-linked agammaglobulinemia type 1, which is an immunodeficiency characterized by the failure to produce mature B lymphocytes, and associated with a failure of Ig heavy chain rearrangement.

Type: Primary
Antigen: BTK
Clonality: Monoclonal
Clone: 7F12H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primates
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR