Anti-GSK3A Mouse Monoclonal Antibody [clone: 9D5G1]

Lieferant: Thermo Fisher Scientific
PIEAMA515338EA 0 EUR
PIEAMA515338
Anti-GSK3A Mouse Monoclonal Antibody [clone: 9D5G1]
Antikörper
MA5-15338 targets GSK3 alpha in ICC and WB applications and shows reactivity with Human samples. The MA5-15338 immunogen is purified recombinant fragment of GSK3 alpha expressed in E. Coli. Glycogen synthase kinase 3-alpha is a multifunctional protein serine kinase, homologous to Drosophila 'shaggy' and implicated in the control of several regulatory proteins including glycogen synthase and transcription factors and PI3K signaling pathways }).

Type: Primary
Antigen: GSK3A
Clonality: Monoclonal
Clone: 9D5G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR