Anti-K5 Mouse Monoclonal Antibody [clone: 7H8C4]

Lieferant: Thermo Fisher Scientific
PIEAMA515348EA 0 EUR
PIEAMA515348
Anti-K5 Mouse Monoclonal Antibody [clone: 7H8C4]
Antikörper
MA5-15348 targets Cytokeratin (Pan) in IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15348 immunogen is purified recombinant fragment of CK5 expressed in E. Coli. The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.

Type: Primary
Antigen: K5
Clonality: Monoclonal
Clone: 7H8C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR