Anti-53BP1 Rabbit Polyclonal Antibody

Lieferant: Thermo Fisher Scientific
PIEAPA116565EA 0 EUR
PIEAPA116565
Anti-53BP1 Rabbit Polyclonal Antibody
Antikörper
Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Suggested positive control: U205, 293T, MO59K cell lysates and MEF lysates . P53-binding protein 1 (53BP1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, it is phosphorylated and relocalizes to the presumptive sites of damage, specifically, double-strand breaks. This also suggests a role in DNA repair, maintaining genomic stability.

Type: Primary
Antigen: 53BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat, Human, Fish, Mouse, Primates, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR