|
![]() |
BESTELLEN
TECHNISCHE DATEN
Antigen symbol | CTR1 |
Antigen name | Copper Transporter 1 |
Reaktivität | Mouse, Rat, Porcine, Human, Xenopus laevis, Zebrafish |
Konjugation | Unconjugated |
Klonalität | Polyclonal |
Antikörper Typ | Primary |
Wirt | Rabbit |
Isotyp | IgG |
Western blot | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Antigen synonyms | COPT1, CTR1, copper transporter 1, solute carrier family 31 (copper tansporters) member 1, hCTR1 |
Gene ID | 171135, 397186, 1317, 403028, 20529, 379422 |
Konzentration | 1 mg/ml |
Immunogen | Synthetic peptide of C-terminal region of human CTR1. |
Purification | Antigen affinity chromatography |
Form | Liquid |
Storage buffer | tris phosphate, tris citrate, 0.1% sodium azide, pH 7-8 |
Lagertemperatur | 4° C, do not freeze |
Transporttemperatur | Ice |