Anti-Phospho-CREB (Ser121) Rabbit Polyclonal Antibody

Lieferant: Thermo Fisher Scientific
PIEAPA116589EA 0 EUR
PIEAPA116589
Anti-Phospho-CREB (Ser121) Rabbit Polyclonal Antibody
Antikörper
Suggested positive control: antigen standard for CREB1 (transient overexpression lysate). Cyclic AMP Response Element Binding protein (CREB) is a basic / leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. CREB is also regulated by DNA-damaging stimuli and is directly phosphorylated on Ser-121 by the ataxia-telangiectasia-mutated protein kinase, ATM in response to ionizing radiation.

Type: Primary
Antigen: Phospho-CREB (Ser121)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR