Anti-GST tag Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: GenScript Biotech
GENSA01380-40EA 164 EUR
GENSA01380-40
Anti-GST tag Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Type: Tag
Antigen: GST tag (Glutathione-S-Transferase tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen