Anti-PGC-1, C-Terminal (777-797) Rabbit Polyclonal Antibody
Lieferant: Oncogene
516557-100EA 511 EUR
516557-100
Anti-PGC-1, C-Terminal (777-797) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PGC-1, C-Terminal (777-797)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen