Anti-P-Glycoprotein (C219) Mouse Monoclonal Antibody [clone: C219]

Lieferant: Oncogene
517310-500EA 560 EUR
517310-500
Anti-P-Glycoprotein (C219) Mouse Monoclonal Antibody [clone: C219]
Type: Primary
Antigen: P-Glycoprotein (C219)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR