Anti-Phosphotyrosine Rabbit Polyclonal Antibody
Lieferant: Oncogene
525290-100EA 444 EUR
525290-100
Anti-Phosphotyrosine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Phosphotyrosine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan