Anti-19S Proteasome S2-Subunit Rabbit Polyclonal Antibody
Lieferant: Oncogene
539166-100EA 446 EUR
539166-100
Anti-19S Proteasome S2-Subunit Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 19S Proteasome S2-Subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human