Anti-PP2A/A (7-19) (PR65) Rabbit Polyclonal Antibody
Lieferant: Oncogene
539509-100EA 571 EUR
539509-100
Anti-PP2A/A (7-19) (PR65) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PP2A/A (7-19) (PR65)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen