Anti-Rab9 (mab9) Mouse Monoclonal Antibody [clone: mab9]
Lieferant: Oncogene
552101-100EA 492 EUR
552101-100
Anti-Rab9 (mab9) Mouse Monoclonal Antibody [clone: mab9]
Antikörper
Type: Primary
Antigen: Rab9 (mab9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen