Anti-MAPK9 Rabbit Polyclonal Antibody

Lieferant: Oncogene
559309-10EA 640 EUR
559309-10
Anti-MAPK9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR