Anti-SHC1 Mouse Monoclonal Antibody [clone: 6E10]

Lieferant: Oncogene
566807-1EA 476 EUR
566807-1
Anti-SHC1 Mouse Monoclonal Antibody [clone: 6E10]
Antikörper
Type: Primary
Antigen: SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)
Clonality: monoclonal
Clone: 60000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR