Anti-SRC Mouse Monoclonal Antibody [clone: 9A6]
Lieferant: Oncogene
569372-100EA 514 EUR
569372-100
Anti-SRC Mouse Monoclonal Antibody [clone: 9A6]
Antikörper
Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: monoclonal
Clone: 9A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen