Anti-SYN1 Rabbit Polyclonal Antibody

Lieferant: Oncogene
574778-50EA 194 EUR
574778-50
Anti-SYN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SYN1 (synapsin I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR