Anti-Syntaxin 4 (2-23) Rabbit Polyclonal Antibody

Lieferant: Oncogene
574788-1EA 324 EUR
574788-1
Anti-Syntaxin 4 (2-23) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Syntaxin 4 (2-23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR