Anti-Telomerase Rabbit Polyclonal Antibody

Lieferant: Oncogene
582000-100EA 529 EUR
582000-100
Anti-Telomerase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Telomerase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR