Anti-Trypsin Rabbit Polyclonal Antibody

Lieferant: Oncogene
650297-1EA 612 EUR
650297-1
Anti-Trypsin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Trypsin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR