Anti-TPSAB1 Rabbit Polyclonal Antibody

Lieferant: Oncogene
650367-500EA 508 EUR
650367-500
Anti-TPSAB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPSAB1 (tryptase alpha/beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR