Anti-TNF Rabbit Polyclonal Antibody

Lieferant: Oncogene
654250-1EA 573 EUR
654250-1
Anti-TNF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR