Anti-UCP1 Rabbit Polyclonal Antibody

Lieferant: Oncogene
662045-100EA 514 EUR
662045-100
Anti-UCP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UCP1 (uncoupling protein 1 (mitochondrial, proton carrier))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR