Anti-Ca V 2.2 (851-867) Rabbit Polyclonal Antibody

Lieferant: Oncogene
681505-1EA 320 EUR
681505-1
Anti-Ca V 2.2 (851-867) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Ca V 2.2 (851-867)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR