Anti-A20 Mouse Monoclonal Antibody [clone: 59A426]

Lieferant: Oncogene
AM63-100EA 492 EUR
AM63-100
Anti-A20 Mouse Monoclonal Antibody [clone: 59A426]
Antikörper
Type: Primary
Antigen: A20
Clonality: monoclonal
Clone: 59A426
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR