Anti-TBK1 (108A429) Mouse Monoclonal Antibody [clone: 108A429]
Lieferant: Oncogene
AM76-100EA 492 EUR
AM76-100
Anti-TBK1 (108A429) Mouse Monoclonal Antibody [clone: 108A429]
Antikörper
Type: Primary
Antigen: TBK1 (108A429)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human