Anti-GAPDH (6C5) Mouse Monoclonal Antibody

Lieferant: Oncogene
CB1001-500EA 444 EUR
CB1001-500
Anti-GAPDH (6C5) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GAPDH (6C5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR